Please login to your online account to display your discounted pricing

Alfa Aesar™ (Cyanomethyl)triphenylphosphonium chloride, 98+%

$92.66 - $92.66

Chemical Identifiers

Chemical Name or Material (Cyanomethyl)triphenylphosphonium chloride
CAS 4336-70-3
Molecular Formula NCCH2P(C6H5)3Cl
Formula Weight 337.79
MDL Number MFCD00031672
UN Number UN3439
Beilstein 6222047
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1309614

 
alfa aesar™
A1309614
25g Each for $92.66

AAA1309606

 
alfa aesar™
A1309606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1309622

 
alfa aesar™
A1309622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1309636

 
alfa aesar™
A1309636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 337.79
Chemical Name or Material (Cyanomethyl)triphenylphosphonium chloride
Assay ≥98%
Molecular Formula NCCH2P(C6H5)3Cl
MDL Number MFCD00031672
CAS 4336-70-3
UN Number UN3439
Beilstein 6222047
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECS Number TA2075000
Melting Point ≈270°C (dec.)
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.