Please login to your online account to display your discounted pricing

Cyanogen bromide, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cyanogen bromide
CAS 506-68-3
Molecular Formula BrCN
Formula Weight 105.93
MDL Number MFCD00011597
UN Number UN1889
Merck Index 14,2693
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3273122

 
alfa aesar
3273122
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 105.93
Melting Point 50° to 52°C
Quantity 100g
Chemical Name or Material Cyanogen bromide
TSCA TSCA
Physical Form Crystalline
Molecular Formula BrCN
MDL Number MFCD00011597
CAS 506-68-3
UN Number UN1889
Merck Index 14,2693
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330;
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CYANOGEN BROMIDE
RTECS Number GT2100000
Density 2.015
Odor Acrid
Boiling Point 61.4°C
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330;
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.