missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanogen bromide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 110785000

 View more versions of this product

Catalog No. AC110785000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Cyanogen bromide
White
97%
Crystals
97%
BrCN
MFCD00011597
15, 2683
1.44
500g
506-68-3
≥96.0%
CBrN
105.93
01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127
Bromine cyanide

Chemical Identifiers

506-68-3
CBrN
105.93
Bromine cyanide
≥96.0%
97%
BrCN
MFCD00011597
Cyanogen bromide, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Refrigerator +4°C