Please login to your online account to display your discounted pricing

Cyanoethylesucrose, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  405940100

 View more versions of this product

Catalog No. AC405940100

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Formula Weight 766.81
Quantity 10g
Color Clear pale yellow to yellow
EINECS Number 242-187-3
Density 1.2000g/mL
TSCA TSCA
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Molecular Formula C36H46N8O11
Chemical Name or Material Cyanoethyl sucrose
MDL Number MFCD00051365
CAS 18304-13-7
Water 1.5% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Cyanoethyl sucrose
CAS 18304-13-7
Molecular Formula C36H46N8O11
Formula Weight 766.81
MDL Number MFCD00051365