missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanoethyl sucrose, Acros Organics™

$168.40 - $168.40

Chemical Identifiers

CAS 18304-13-7
Molecular Formula C36H46N8O11
Molecular Weight (g/mol) 766.809
MDL Number MFCD00051365
InChI Key XCKPLVGWGCWOMD-YYEYMFTQSA-N
Synonym cyanoethyl sucrose, octakis-o-2-cyanoethyl sucrose, alpha-d-glucopyranoside, 1,3,4,6-tetrakis-o-2-cyanoethyl-beta-d-fructofuranosyl 2,3,4,6-tetrakis-o-2-cyanoethyl, 1-o,3-o,4-o,6-o-tetrakis 2-cyanoethyl-beta-d-fructofuranosyl 2-o,3-o,4-o,6-o-tetrakis 2-cyanoethyl-alpha-d-glucopyranoside, 3-2r,3r,4s,5r,6r-6-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxymethyl oxolan-2-yl oxy-3,4,5-tris 2-cyanoethoxy oxan-2-yl methoxy propanenitrile, 3-2r,3r,4s,5r,6r-6-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxymethyl tetrahydrofuran-2-yl oxy-3,4,5-tris 2-cyanoethoxy tetrahydropyran-2-yl methoxy propanenitrile, 3-2r,3r,4s,5r,6r-2-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxy methyl oxolan-2-yl oxy-4,5-bis 2-cyanoethoxy-6-2-cyanoethoxy methyl oxan-3-yl oxy propanenitrile
PubChem CID 87570
IUPAC Name 3-[[(2R,3R,4S,5R,6R)-6-[(2S,3S,4R,5R)-3,4-bis(2-cyanoethoxy)-2,5-bis(2-cyanoethoxymethyl)oxolan-2-yl]oxy-3,4,5-tris(2-cyanoethoxy)oxan-2-yl]methoxy]propanenitrile
SMILES C(COCC1C(C(C(C(O1)OC2(C(C(C(O2)COCCC#N)OCCC#N)OCCC#N)COCCC#N)OCCC#N)OCCC#N)OCCC#N)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC405940100 SDS View Documents Acros Organics
405940100
10g Glass bottle Each for $168.40
Specifications

Chemical Identifiers

18304-13-7
766.809
XCKPLVGWGCWOMD-YYEYMFTQSA-N
87570
C(COCC1C(C(C(C(O1)OC2(C(C(C(O2)COCCC#N)OCCC#N)OCCC#N)COCCC#N)OCCC#N)OCCC#N)OCCC#N)C#N
C36H46N8O11
MFCD00051365
cyanoethyl sucrose, octakis-o-2-cyanoethyl sucrose, alpha-d-glucopyranoside, 1,3,4,6-tetrakis-o-2-cyanoethyl-beta-d-fructofuranosyl 2,3,4,6-tetrakis-o-2-cyanoethyl, 1-o,3-o,4-o,6-o-tetrakis 2-cyanoethyl-beta-d-fructofuranosyl 2-o,3-o,4-o,6-o-tetrakis 2-cyanoethyl-alpha-d-glucopyranoside, 3-2r,3r,4s,5r,6r-6-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxymethyl oxolan-2-yl oxy-3,4,5-tris 2-cyanoethoxy oxan-2-yl methoxy propanenitrile, 3-2r,3r,4s,5r,6r-6-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxymethyl tetrahydrofuran-2-yl oxy-3,4,5-tris 2-cyanoethoxy tetrahydropyran-2-yl methoxy propanenitrile, 3-2r,3r,4s,5r,6r-2-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxy methyl oxolan-2-yl oxy-4,5-bis 2-cyanoethoxy-6-2-cyanoethoxy methyl oxan-3-yl oxy propanenitrile
3-[[(2R,3R,4S,5R,6R)-6-[(2S,3S,4R,5R)-3,4-bis(2-cyanoethoxy)-2,5-bis(2-cyanoethoxymethyl)oxolan-2-yl]oxy-3,4,5-tris(2-cyanoethoxy)oxan-2-yl]methoxy]propanenitrile

Specifications

1.2000g/mL
Authentic
1.4830 to 1.4850
Yellow
18304-13-7
MFCD00051365
XCKPLVGWGCWOMD-YYEYMFTQSA-N
3-[[(2R,3R,4S,5R,6R)-6-[(2S,3S,4R,5R)-3,4-bis(2-cyanoethoxy)-2,5-bis(2-cyanoethoxymethyl)oxolan-2-yl]oxy-3,4,5-tris(2-cyanoethoxy)oxan-2-yl]methoxy]propanenitrile
87570
Viscous Liquid
>180°C
Glass bottle
1.5% max. (K.F.)
10g
C36H46N8O11
cyanoethyl sucrose, octakis-o-2-cyanoethyl sucrose, alpha-d-glucopyranoside, 1,3,4,6-tetrakis-o-2-cyanoethyl-beta-d-fructofuranosyl 2,3,4,6-tetrakis-o-2-cyanoethyl, 1-o,3-o,4-o,6-o-tetrakis 2-cyanoethyl-beta-d-fructofuranosyl 2-o,3-o,4-o,6-o-tetrakis 2-cyanoethyl-alpha-d-glucopyranoside, 3-2r,3r,4s,5r,6r-6-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxymethyl oxolan-2-yl oxy-3,4,5-tris 2-cyanoethoxy oxan-2-yl methoxy propanenitrile, 3-2r,3r,4s,5r,6r-6-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxymethyl tetrahydrofuran-2-yl oxy-3,4,5-tris 2-cyanoethoxy tetrahydropyran-2-yl methoxy propanenitrile, 3-2r,3r,4s,5r,6r-2-2s,3s,4r,5r-3,4-bis 2-cyanoethoxy-2,5-bis 2-cyanoethoxy methyl oxolan-2-yl oxy-4,5-bis 2-cyanoethoxy-6-2-cyanoethoxy methyl oxan-3-yl oxy propanenitrile
C(COCC1C(C(C(C(O1)OC2(C(C(C(O2)COCCC#N)OCCC#N)OCCC#N)COCCC#N)OCCC#N)OCCC#N)OCCC#N)C#N
766.809
766.81
Cyanoethyl sucrose
Safety and Handling

Safety and Handling

EINECSNumber : 242-187-3

TSCA : TSCA