missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanoacetylurea, TCI America™

$44.31 - $261.25

Chemical Identifiers

CAS 1448-98-2
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00007947
InChI Key QJGRPCPCQQPZLZ-UHFFFAOYSA-N
Synonym cyanoacetylurea, 2-cyanoacetylurea, acetamide, n-aminocarbonyl-2-cyano, cyanoacetyl urea, urea, cyanoacetyl, 2-cyanoacetyl urea, amino-n-2-cyanoacetyl amide, cyanoacetyl urea, n-2-cyanoacetyl urea, n-cyanoacetyl urea
PubChem CID 74055
IUPAC Name N-carbamoyl-2-cyanoacetamide
SMILES C(C#N)C(=O)NC(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C110125G View Documents TCI America
C110125G
25g Each for $44.31
 
C1101250G View Documents TCI America
C1101250G
250g Each for $261.25
 
Specifications

Chemical Identifiers

1448-98-2
127.103
QJGRPCPCQQPZLZ-UHFFFAOYSA-N
74055
C(C#N)C(=O)NC(=O)N
C4H5N3O2
MFCD00007947
cyanoacetylurea, 2-cyanoacetylurea, acetamide, n-aminocarbonyl-2-cyano, cyanoacetyl urea, urea, cyanoacetyl, 2-cyanoacetyl urea, amino-n-2-cyanoacetyl amide, cyanoacetyl urea, n-2-cyanoacetyl urea, n-cyanoacetyl urea
N-carbamoyl-2-cyanoacetamide

Specifications

White-Yellow
1448-98-2
MFCD00007947
cyanoacetylurea, 2-cyanoacetylurea, acetamide, n-aminocarbonyl-2-cyano, cyanoacetyl urea, urea, cyanoacetyl, 2-cyanoacetyl urea, amino-n-2-cyanoacetyl amide, cyanoacetyl urea, n-2-cyanoacetyl urea, n-cyanoacetyl urea
C(C#N)C(=O)NC(=O)N
127.103
127.10
≥98.0% (T)
25g
C4H5N3O2
3439
QJGRPCPCQQPZLZ-UHFFFAOYSA-N
N-carbamoyl-2-cyanoacetamide
74055
Crystal-Powder at 20°C
Cyanoacetylurea
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3324

TSCA : Yes