Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyanoacetylurea, 97%

$50.18 - $541.82

Chemical Identifiers

Chemical Name or Material Cyanoacetylurea
CAS 1448-98-2
Molecular Formula NCCH2CONHCONH2
Formula Weight 127.1
MDL Number MFCD00007947
Beilstein 1705616
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB209950B

 
alfa aesar™
B209950B
1000g Each for $541.82

AAB2099518

 
alfa aesar™
B2099518
50g Each for $50.18

AAB2099530

 
alfa aesar™
B2099530
250g Each for $164.70
Description & Specifications

Specifications

Formula Weight 127.1
TSCA TSCA
Assay 97%
Molecular Formula NCCH2CONHCONH2
Chemical Name or Material Cyanoacetylurea
MDL Number MFCD00007947
CAS 1448-98-2
Beilstein 1705616
Health Hazard 1 Danger
Melting Point ≈215°C (dec.)
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1000g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.