Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyanoacetic acid hydrazide, 98%

Chemical Identifiers

Chemical Name or Material Cyanoacetic acid hydrazide
CAS 140-87-4
Molecular Formula C3H5N3O
Linear Formula NCCH2CONHNH2
Formula Weight 99.09
MDL Number MFCD00007611
UN Number UN2811
Beilstein 1751498
Merck Index 14,2680
Synonym Cyacetacide; Cyanoacetohydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2437014

 
alfa aesar™
B2437014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2437022

 
alfa aesar™
B2437022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 99.09
Chemical Name or Material Cyanoacetic acid hydrazide
Assay 98%
Molecular Formula C3H5N3O
Linear Formula NCCH2CONHNH2
MDL Number MFCD00007611
CAS 140-87-4
UN Number UN2811
Beilstein 1751498
Merck Index 14,2680
Synonym Cyacetacide; Cyanoacetohydrazide
Health Hazard 1 Danger
Melting Point 105° to 110°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number AG4200000
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C