Learn More
Cyanoacetic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A110450B
Description
Cyanoacetic acid is used in pharmaceutical industry for producing vitamin B6 and caffeine. It is also used in making dyes, agricultural chemicals, and in the synthesis of phenylacetic acid esters. It acts as a precursor cyanoacrylates viz. ethyl cyanoacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyanoacetic acid is used in pharmaceutical industry for producing vitamin B6 and caffeine. It is also used in making dyes, agricultural chemicals, and in the synthesis of phenylacetic acid esters. It acts as a precursor cyanoacrylates viz. ethyl cyanoacrylate.
Solubility
Miscible with water, ethyl alcohol, ether. Slightly miscible with benzene, acetic acid and chloroform.
Notes
Hygroscopic. Incompatible with strong bases, acids, strong oxidizing agents, alkaline, nitriles and reducing agents.
Specifications
Cyanoacetic acid | |
64°C to 69°C | |
108°C (15 mmHg) | |
Odorless | |
MFCD00002677 | |
UN3261 | |
Hygroscopic | |
cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova | |
MLIREBYILWEBDM-UHFFFAOYSA-N | |
2-cyanoacetic acid | |
9740 | |
85.06 |
372-09-8 | |
1.5 | |
107°C (224°F) | |
C3H3NO2 | |
1000 g | |
506325 | |
14,2689 | |
Miscible with water,ethyl alcohol,ether. Slightly miscible with benzene,acetic acid and chloroform. | |
C(C#N)C(=O)O | |
85.062 | |
CHEBI:51889 | |
99% |
Chemical Identifiers
372-09-8 | |
85.062 | |
MLIREBYILWEBDM-UHFFFAOYSA-N | |
9740 | |
2-cyanoacetic acid |
C3H3NO2 | |
MFCD00002677 | |
cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova | |
CHEBI:51889 | |
C(C#N)C(=O)O |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-743-9
RTECSNumber : AG3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only