Please login to your online account to display your discounted pricing

α-Cyano-4-hydroxycinnamic acid, 97%, ACROS Organics™

$46.09 - $46.09

Chemical Identifiers

Chemical Name or Material α-Cyano-4-hydroxycinnamic acid
CAS 28166-41-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H7NO3
Linear Formula HOC6H4CH=C(CN)CO2H
Formula Weight 189.17
MDL Number MFCD00004204
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC163440050

  SDS
thermo fisher scientific
163440050
Glass bottle 5g Each for $46.09
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 189.17
Quantity 5g
Color Yellow
Beilstein 10, 520
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material α-Cyano-4-hydroxycinnamic acid
Assay Percent Range 97%
Molecular Formula C10H7NO3
CAS 28166-41-8
Infrared Spectrum Authentic
Linear Formula HOC6H4CH=C(CN)CO2H
MDL Number MFCD00004204
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.