Please login to your online account to display your discounted pricing

α-Cyano-3-hydroxycinnamic acid, 99%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  163090250

 View more versions of this product

Catalog No. AC163090250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 189.17
Physical Form Crystalline Powder
Quantity 25g
Color Green-yellow
TSCA TSCA
Melting Point 230° to 233°C
EINECS Number 259-289-9
Beilstein 10, 520
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula HOC6H4CH=C(CN)CO2H
Assay Percent Range 99%
CAS 54673-07-3
CAS Min % 98.5
Name Note 99%
Chemical Name or Material α-Cyano-3-hydroxycinnamic acid
Molecular Formula C10H7NO3
MDL Number MFCD00004203
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

Specifications

Chemical Name or Material α-Cyano-3-hydroxycinnamic acid
Name Note 99%
CAS 54673-07-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H7NO3
Linear Formula HOC6H4CH=C(CN)CO2H
Formula Weight 189.17
MDL Number MFCD00004203
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

SDS & Safety Documents