missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Cyano-3-hydroxycinnamic acid, 98%, Acros Organics™

Manufacturer:  Acros Organics 163090050

 View more versions of this product

Catalog No. AC163090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

α-Cyano-3-hydroxycinnamic acid
Glass bottle
230.0°C to 233.0°C
98%
98.5
98%
HOC6H4CH=C(CN)CO2H
10, 520
HPLNTJVXWMJLNJ-YWEYNIOJSA-N
(Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid
21338270
Crystalline Powder
Authentic
Green-Yellow
5g
54673-07-3
100.0
C10H7NO3
MFCD00004203
2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O
189.17
189.17
98%

Chemical Identifiers

54673-07-3
189.17
HPLNTJVXWMJLNJ-YWEYNIOJSA-N
21338270
C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O
C10H7NO3
MFCD00004203
2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
(Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-289-9

TSCA : TSCA

SDS Documents