missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Cyano-3-hydroxycinnamic acid, 98%, Acros Organics™

Chemical Identifiers

CAS 54673-07-3
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00004203
InChI Key HPLNTJVXWMJLNJ-YWEYNIOJSA-N
Synonym 2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
PubChem CID 21338270
IUPAC Name (Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC163090250 SDS View Documents Acros Organics
163090250
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163090050 SDS View Documents Acros Organics
163090050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163091000 SDS View Documents Acros Organics
163091000
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

54673-07-3
189.17
HPLNTJVXWMJLNJ-YWEYNIOJSA-N
21338270
C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O
C10H7NO3
MFCD00004203
2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
(Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid

Specifications

Authentic
Green-Yellow
25g
98.5
98%
HOC6H4CH=C(CN)CO2H
10, 520
HPLNTJVXWMJLNJ-YWEYNIOJSA-N
(Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid
21338270
Crystalline Powder
α-Cyano-3-hydroxycinnamic acid, 98%
Glass bottle
230.0°C to 233.0°C
54673-07-3
100.0
C10H7NO3
MFCD00004203
2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O
189.17
189.17
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-289-9

TSCA : TSCA