missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanide, Standard solution, Specpure, CN 1000μg/mL, Alfa Aesar™

Manufacturer:  Alfa Aesar 43804AE

 View more versions of this product

Catalog No. AA43804AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyanide
KCN in 0.1% NaOH
CN: 1000μg/mL
Odorless
Colorless
Standard Solution
MFCD00468042
Liquid
1g/cm3 at 20°C
100mL

Chemical Identifiers

KCN in 0.1% NaOH
MFCD00468042
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Wear suitable protective clothing and gloves.

H301-H311-H332: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled.

Documents