missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanide Standard, Certified, 1000ppm ±10ppm (1mL = 1mg CN-), LabChem™

$30.00 - $90.91

Chemical Identifiers

CAS 7732-18-5,151-50-8,1310-73-2
CAS Max % 99.55,0.25,0.2
Linear Formula KCN
Formula Weight 65.12
Chemical Name or Material Cyanide Standard, 1000ppm (1mL = 1mg CN-)
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC135451 Lab Chem
LC135451
Poly Bottle 500mL Each for $66.31
LC135452 Lab Chem
LC135452
Poly Bottle 1L Each for $90.91
LC135457 Lab Chem
LC135457
Poly Bottle 125mL Each for $30.00
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,151-50-8,1310-73-2
KCN
Cyanide Standard, 1000ppm (1mL = 1mg CN-)
99.55,0.25,0.2
65.12
Certified Grade

Specifications

7732-18-5,151-50-8,1310-73-2
Liquid
KCN
Passes Test
Soluble in water
1g/mL
500mL
Certified Grade
99.55,0.25,0.2
1000ppm ±10ppm (1mL = 1mg CN-)
65.12
Poly Bottle
Traceable to NIST
Clear, colorless
Cyanide Standard, 1000ppm (1mL = 1mg CN-)

Safety and Handling

GHS H Statement
Harmful to aquatic life.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

Recommended Storage : Room Temperature