missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cy-cBRIDP(regR), TCI America™

Manufacturer:  TCI America C2980200MG

 View more versions of this product

Catalog No. C2980200MG



Specifications

Specifications

Cy-cBRIDP(regR)
122°C
1023330-38-2
MFCD10565626
JDNAVZKVTWHWKB-UHFFFAOYSA-N
dicyclohexyl-(1-methyl-2,2-diphenylcyclopropyl)phosphane
53428829
Crystal-Powder at 20°C
White-Yellow
200mg
C28H37P
Dicyclohexyl(1-methyl-2,2-diphenylcyclopropyl)phosphine
CC1(CC1(C2=CC=CC=C2)C3=CC=CC=C3)P(C4CCCCC4)C5CCCCC5
404.578
404.58
≥95.0% (HPLC)

Chemical Identifiers

1023330-38-2
404.578
JDNAVZKVTWHWKB-UHFFFAOYSA-N
53428829
CC1(CC1(C2=CC=CC=C2)C3=CC=CC=C3)P(C4CCCCC4)C5CCCCC5
C28H37P
MFCD10565626
Dicyclohexyl(1-methyl-2,2-diphenylcyclopropyl)phosphine
dicyclohexyl-(1-methyl-2,2-diphenylcyclopropyl)phosphane
Safety and Handling

Safety and Handling

TSCA : No

Documents