CY 208-243, Tocris
Selective D1-like agonist
Manufacturer:  R & D Systems 124950
 View more versions of this product
Catalog No. 124950
Specifications Description & Specifications
Specifications
CY 208-243
100999-26-6
C19H18N2
(-)-(6aR,12bR)-4,6,6a,7,8,12b-Hexahydro-7-methylindolo[4,3-a]phenanthridin
50mg
>98%
274.36
Chemical Identifiers
>98%
274.36
CY 208-243
C19H18N2
(-)-(6aR,12bR)-4,6,6a,7,8,12b-Hexahydro-7-methylindolo[4,3-a]phenanthridin
Safety and Handling
Recommended Storage : Store at Room Temperature