Please login to your online account to display your discounted pricing

CY 208-243, Tocris


Selective D1-like agonist

Quantity 10mg

Manufacturer: r & d systems  124910

 View more versions of this product

Catalog No. 124910

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material CY 208-243
Quantity 10mg
CAS 100999-26-6
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C19H18N2
Formula Weight 274.36
Synonym (-)-(6aR,12bR)-4,6,6a,7,8,12b-Hexahydro-7-methylindolo[4,3-a]phenanthridin

Specifications

Chemical Name or Material CY 208-243
Assay Percent Range >98%
Molecular Formula C19H18N2
Formula Weight 274.36
Synonym (-)-(6aR,12bR)-4,6,6a,7,8,12b-Hexahydro-7-methylindolo[4,3-a]phenanthridin
Accessories

Store at Room Temperature