Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in western blotting and flow cytometry experiments with Thermo Scientific™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody.

$521.94 - $1,718.65

Specifications

Antigen CXCR4/Fusin/LESTR/HUMSTR Ab-2
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1615P

 
thermo scientific™ lab vision™
RB1615P
Purified Ab with BSA and Azide 1mL Each for $1,563.53

RB1615P0

 
thermo scientific™ lab vision™
RB1615P0
Purified Ab with BSA and Azide 100μL Each for $521.94

RB1615P1

 
thermo scientific™ lab vision™
RB1615P1
Purified Ab with BSA and Azide 500μL Each for $1,093.37

RB1615P1ABX

 
thermo scientific™ lab vision™
RB1615P1ABX
Ab WWithout BSA/Azide (Frozen) 500μL Each for $1,193.88

RB1615PABX

 
thermo scientific™ lab vision™
RB1615PABX
Ab Without BSA/Azide (Frozen) 1mL Each for $1,718.65
Description & Specifications

Specifications

Antigen CXCR4/Fusin/LESTR/HUMSTR Ab-2
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline G Protein-Coupled Receptors
Type Primary
Immunogen A synthetic peptide derived from the second extracellular loop of human CXCR4
Isotype IgG

Ensure accurate, reproducible results in western blotting and flow cytometry experiments with Thermo Scientific™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody.

CXCR4, also termed Fusin, LESTR, or HUMSTR is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CXCR4 is expressed predominantly on nanve T cell subset of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. It is also expressed on neutrophils, monocytes and various cell lines.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey and Mouse. Others not known.

Epitope: Extracellular

Immunogen: A synthetic peptide derived from the second extracellular loop of human CXCR4

Molecular Weight: 40 kDa

Positive Control: Raji cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO