Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in western blotting and flow cytometry experiments with Thermo Scientific™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1615PABX

 View more versions of this product

Catalog No. RB1615PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Antigen CXCR4/Fusin/LESTR/HUMSTR Ab-2
Host Species Rabbit
Applications Western Blotting
Isotype IgG
Immunogen A synthetic peptide derived from the second extracellular loop of human CXCR4
Format Concentrated
Quantity 1mL
Concentration 1mg/mL
Conjugate Unlabeled
Type Primary
Formulation Ab Without BSA/Azide (Frozen)
Research Discipline G Protein-Coupled Receptors
Classification Polyclonal

Ensure accurate, reproducible results in western blotting and flow cytometry experiments with Thermo Scientific™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody.

CXCR4, also termed Fusin, LESTR, or HUMSTR is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CXCR4 is expressed predominantly on nanve T cell subset of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. It is also expressed on neutrophils, monocytes and various cell lines.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey and Mouse. Others not known.

Epitope: Extracellular

Immunogen: A synthetic peptide derived from the second extracellular loop of human CXCR4

Molecular Weight: 40 kDa

Positive Control: Raji cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO