Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in western blotting and flow cytometry experiments with Thermo Scientific™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1615P1ABX

 View more versions of this product

Catalog No. RB1615P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Isotype IgG
Host Species Rabbit
Antigen CXCR4/Fusin/LESTR/HUMSTR Ab-2
Immunogen A synthetic peptide derived from the second extracellular loop of human CXCR4
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 500µL
Format Concentrated
Regulatory Status RUO
Conjugate Unlabeled
Type Primary
Formulation Ab WWithout BSA/Azide (Frozen)
Concentration 1mg/mL
Classification Polyclonal
Research Discipline G Protein-Coupled Receptors

Ensure accurate, reproducible results in western blotting and flow cytometry experiments with Thermo Scientific™ CXCR4/Fusin/LESTR/HUMSTR Ab-2, Rabbit Polyclonal Antibody.

CXCR4, also termed Fusin, LESTR, or HUMSTR is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CXCR4 is expressed predominantly on nanve T cell subset of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. It is also expressed on neutrophils, monocytes and various cell lines.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey and Mouse. Others not known.

Epitope: Extracellular

Immunogen: A synthetic peptide derived from the second extracellular loop of human CXCR4

Molecular Weight: 40 kDa

Positive Control: Raji cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO