Please login to your online account to display your discounted pricing

CXCL9 (MIG) Armenian Hamster anti-Mouse, PE, Clone: MIG-2F5.5, eBioscience™

Armenian Hamster Monoclonal Antibody

$187.05 - $506.05

Specifications

Antigen CXCL9 (MIG)
Clone MIG-2F5.5
Host Species Armenian Hamster
Gene Alias monokine induced by IFN-g
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010746

 
life technologies
LS12300980
25μg Each for $187.05

5010747

 
life technologies
LS12300982
100μg Each for $506.05
Description & Specifications

Specifications

Antigen CXCL9 (MIG)
Clone MIG-2F5.5
Host Species Armenian Hamster
Gene Alias monokine induced by IFN-g
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody MIG-2F5.5 reacts with mouse monokine induced by IFN gamma (mig, mig-1, CXCL9). This 14.4kDa inflammatory chemokine is specifically induced by IFN gamma, but not other activators such as LPS or IFN alpha. Secretion of mig, mainly by macrophages results in the chemotaxis of a variety of activated T cells via the CXCR3 chemokine receptor. Mig is involved many areas of research including autoimmune diseases, cancer, and inflammation.