Please login to your online account to display your discounted pricing

CXCL10 (IP-10) Mouse anti-Human, PerCP-eFluor 710, Clone: 4NY8UN, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $448.05

Specifications

Antigen CXCL10 (IP-10)
Clone 4NY8UN
Host Species Murine
Gene Alias Interferon Inducible Protein-10, CXC Motif Chemokine 10
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123959

 
life technologies
LS46974441
25 Tests Each for $187.05

501123960

 
life technologies
LS46974442
100 Tests Each for $448.05
Description & Specifications

Specifications

Antigen CXCL10 (IP-10)
Clone 4NY8UN
Host Species Murine
Gene Alias Interferon Inducible Protein-10, CXC Motif Chemokine 10
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This 4NY8UN monoclonal antibody reacts with human CXCL10. CXCL10, also known as Interferon Inducible Protein-10 (IP-10), is a member of the CXC family of chemokines. It is secreted by monocytes, epithelial cells, and endothelial cells in response to IFN gamma or other pro-inflammatory cytokines and stimuli. CXCL10 signaling is mediated by the G protein-coupled receptor CXCR3, which is expressed on activated T cells and plays an important role in directing the migration of T cells, especially during Th1 responses. In addition to its chemotactic functions, CXCL10 is a potent inhibitor of angiogenesis and tumor growth and modulates expression of adhesion molecules. Elevated levels of CXCL10 are observed in response to allergic inflammation, various infections, and Alzheimer’s disease.