missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CX3CR1 Rat anti-Human, PE, Clone: 2A9-1, eBioscience™

Rat Monoclonal Antibody

$157.08 - $407.88

Specifications

Antigen CX3CR1
Clone 2A9-1
Host Species Rat
Gene Alias Chemokine C-X3-C receptor 1, V28
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245511 Life Technologies
LS12609941
25 Tests Each for $157.08
50245512 Life Technologies
LS12609942
100 Tests Each for $407.88
Specifications

Description

This 2A9-1 monoclonal antibody reacts with human CX3CR1, which is the receptor for fractalkine, a transmembrane chemokine of the CX3C family. CX3CR1 is a seven transmembrane G protein-coupled receptor expressed on CD16+ NK cells, T cells (e.g. CD8+, CD4+, and gamma/delta), and monocytes. In non-immune cells, CX3CR1 has been found on osteoclast precursors and microglia. Little to no CX3CR1 surface expression can be detected on B cells and granulocytes. Together, fractalkine and its receptor mediate cell-cell adhesion and chemotaxis of NK cells, T cells, and monocytes. The expression of CX3CR1 has also been correlated with high levels of intracellular perforin and granzyme B.
Description & Specifications

Specifications

CX3CR1
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
2A9-1
Chemokine C-X3-C receptor 1, V28
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer