Please login to your online account to display your discounted pricing

CX3CR1 Rat anti-Human, Biotin, Clone: 2A9-1, eBioscience™

Rat Monoclonal Antibody

$201.55 - $404.55

Specifications

Antigen CX3CR1
Clone 2A9-1
Host Species Rat
Gene Alias Chemokine C-X3-C receptor 1, V28
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012131

 
life technologies
LS13609980
25μg Each for $201.55

5012132

 
life technologies
LS13609982
100μg Each for $404.55
Description & Specifications

Specifications

Antigen CX3CR1
Clone 2A9-1
Host Species Rat
Gene Alias Chemokine C-X3-C receptor 1, V28
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This 2A9-1 monoclonal antibody reacts with human CX3CR1, which is the receptor for fractalkine, a transmembrane chemokine of the CX3C family. CX3CR1 is a seven transmembrane G protein-coupled receptor expressed on CD16+ NK cells, T cells (e.g. CD8+, CD4+, and gamma/delta), and monocytes. In non-immune cells, CX3CR1 has been found on osteoclast precursors and microglia. Little to no CX3CR1 surface expression can be detected on B cells and granulocytes. Together, fractalkine and its receptor mediate cell-cell adhesion and chemotaxis of NK cells, T cells, and monocytes. The expression of CX3CR1 has also been correlated with high levels of intracellular perforin and granzyme B.