missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ pH Gradient Buffers

Robust reproducible gradients that are applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

Manufacturer:  Thermo Scientific™ 085349

 View more versions of this product

Catalog No. DX085349


 

Specifications

Description

Delivers robust reproducible gradients with the Thermo Scientific™ pH buffer platform that is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

The pH gradient platform saves time in method development and facilitates method transfer to QA/QC for a wide range of MAb charge variants through a generic LC-based approach to MAb characterization.

pH buffer solutions can be used to generate highly reproducible, linear pH gradients in cation exchange chromatography.

 

  • Robust, reproducible gradients
  • Predict biomolecule pI
  • Simple method optimization
  • Easily automated
  • Applicable to the majority of MAbs


Unlike traditional cation exchange chromatography using salt gradients, it is possible to predict the pI and the expected retention of the charge variants and use a narrow pH range to get a higher resolution separation.

Description & Specifications

Specifications

CX-1 pH Gradient Buffer Kit
Buffer A (250mL) and Buffer B (250mL)
250mL
5.6 to 10.2