Please login to your online account to display your discounted pricing

Cutaneous Lymphocyte Antigen Rat anti-Human, eFluor 660, Clone: HECA-452, eBioscience™

Rat Monoclonal Antibody

$131.95 - $333.50

Specifications

Antigen Cutaneous Lymphocyte Antigen
Clone HECA-452
Host Species Rat
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124562

 
life technologies
LS50985780
25μg Each for $131.95

501124563

 
life technologies
LS50985782
100μg Each for $333.50
Description & Specifications

Specifications

Antigen Cutaneous Lymphocyte Antigen
Clone HECA-452
Host Species Rat
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blot
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This HECA-452 monoclonal antibody reacts with human Cutaneous Lymphocyte Antigen (CLA). This cell surface glycoprotein is most highly expressed on T cells in the skin. Nevertheless, CLA can also be detected on some peripheral T and NK cells, as well as monocytes, neutrophils and CD1a+ dendritic cells. Studies have determined that this antibody recognizes sialofucosylated glycans and sialyl Lewis x (sLex). CLA, which binds E-selectin and endothelial cell-leukocyte adhesion molecule 1 (ELAM-1), is involved in T lymphocyte infiltration and adhesion at cutaneous sites of inflammation.

The HECA-452 antibody has been reported to inhibit sLex binding to E-selectin; however, this property is still under debate in the literature.