Current Technologies Saf De-Cap™ Tube De-Capper
Tapered shape fits all tube sizes
Manufacturer: Current Technologies S4001
Description
- Provides good tactual feel, are made of flexible and soft plastic, easy to use
- Blocks splashes of blood and specimen when removing caps; particularly helpful when high pressure builds inside tube due to elevation, air-mail of specimens, etc.
- Minimizes splash when breaking open ampules and glass breakage doesn't rip gloves
- Vertical ridges on the outside and fine grooves on the inside control removal of the cap; adaptable to various tube sizes
- Improves personal safety when removing stoppers from tubes / specimens or breaking open ampules. Minimizes cross-contamination as gloved hands touch equipment, keyboards, switches, etc.
- Helps control cost as glove changes are reduced
Specifications
Clear | |
Plastic |