missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Curcumin, 95% (total curcuminoid content), from Turmeric rhizome, Alfa Aesar™

Manufacturer:  Alfa Aesar B2157309

 View more versions of this product

Catalog No. AAB2157309



Description

Description

Applications
Curcumin is used for boron quantification. It inhibits lipoxygenase and cyclooxygenase. It acts as a dietary supplement and is used in pharmaceutical industry. It acts as a food coloring agent and acts as a food additive.

Solubility
Soluble in acetone, dimethylsufoxide, ethanol and dimethyl formamide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Curcumin
from Turmeric rhizome
0.95
C21H20O6
2306965
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
COC1=C(C=CC(=C1)C=CC(=O)CC(=O)C=CC2=CC(=C(C=C2)O)OC)O
368.385
CHEBI:3962
95%
10g
458-37-7
(total curcuminoid content)
MFCD00008365
14,2673
VFLDPWHFBUODDF-FCXRPNKRSA-N
(1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
969516
368.39

Chemical Identifiers

458-37-7
368.385
VFLDPWHFBUODDF-FCXRPNKRSA-N
969516
(1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
C21H20O6
MFCD00008365
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
CHEBI:3962
COC1=C(C=CC(=C1)C=CC(=O)CC(=O)C=CC2=CC(=C(C=C2)O)OC)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

RTECSNumber : MI5230000

TSCA : TSCA

Documents