Please login to your online account to display your discounted pricing

Cupric Sulfate Pentahydrate (Powder/Technical), Fisher Chemical

$2,599.64 - $3,318.14

Chemical Identifiers

Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula CuO4S · 5 H2O
Formula Weight 249.68
MDL Number MFCD00149681
Synonym Copper (II) Sulfate Pentahydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C490-10

  SDS
fisher scientific company
C49010
Poly Pail 10kg Each for $2,599.64

C490-3

  SDS
fisher scientific company
C4903
Poly Bottle 3kg Case of 4 Each for $3,318.14
Description & Specifications

Specifications

Formula Weight 249.68
Purity Grade Technical Grade
Melting Point 110°C
Quantity 10kg
DOT Information DOT Class 9, : Miscellaneous
Physical Form Solid
Molecular Formula CuO4S · 5 H2O
Color Blue
Vapor Pressure 7.3mmHg at 25°C
pH 3.5 to 4.5
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1
ChemAlert Storage Symbol Blue
Chemical Name or Material Cupric Sulfate Pentahydrate
MDL Number MFCD00149681
Identification Pass Test
Packaging Poly Pail
Synonym Copper (II) Sulfate Pentahydrate
Additional Information Vapor Pressure: 7.3mmHg at 25°C
Health Hazard 2 WARNING!
CAS 7758-99-8
Safety and Handling
 

Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1

WARNING!