Please login to your online account to display your discounted pricing

Cupric Sulfate Pentahydrate (Granular/USP), Fisher Chemical


Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLC494212

 View more versions of this product

Catalog No. C494-212

  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 249.68
Melting Point 110°C
Quantity 2.5kg
DOT Information DOT Class 9, : Miscellaneous
Purity Grade USP
Physical Form Solid
Assay 98.5 to 100.5% (USP)
Molecular Formula CuO4S · 5 H2O
Vapor Pressure 7.3mmHg at 25°C
Color Blue
pH 3.5 to 4.5
Chemical Name or Material Cupric Sulfate Pentahydrate
Loss on Drying 33.0 to 36.5% (USP)
Potassium (K) 0.01% max. (USP)
Assay Percent Range (Dried Basis) 98.5to100.5%
CAS 7758-99-8
Synonym Copper (II) Sulfate Pentahydrate
Calcium (Ca) 0.005% max. (USP)
Identification Pass Test (USP)
Iron (Fe) 0.003% max. (USP)
Nickel (Ni) 0.005% max. (USP)
Sodium (Na) 0.02% max. (USP)
Assay Percent Notes (Dried Basis) 98.5-100.5%
MDL Number MFCD00149681
Packaging Poly Bottle
Residual Solvents Meets Requirements (USP)
Additional Information Vapor Pressure: 7.3mmHg at 25°C
ChemAlert Storage Symbol Blue
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1

Specifications

Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Purity Grade USP
Physical Form Solid
Assay 98.5 to 100.5% (USP)
Assay Percent Range (Dried Basis) 98.5to100.5%
Assay Percent Notes (Dried Basis) 98.5-100.5%
Molecular Formula CuO4S · 5 H2O
Formula Weight 249.68
MDL Number MFCD00149681
Synonym Copper (II) Sulfate Pentahydrate
Accessories
 

Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1

WARNING!

SDS & Safety Documents