Please login to your online account to display your discounted pricing

Cupric Sulfate Pentahydrate (Granular/USP), Fisher Chemical

$235.36 - $1,859.40

Chemical Identifiers

Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Purity Grade USP
Physical Form Solid
Assay 98.5 to 100.5% (USP)
Assay Percent Range (Dried Basis) 98.5to100.5%
Assay Percent Notes (Dried Basis) 98.5-100.5%
Molecular Formula CuO4S · 5 H2O
Formula Weight 249.68
MDL Number MFCD00149681
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C494-212

  SDS
fisher scientific company
FLC494212
Poly Bottle 2.5kg Case of 4 Each for $1,859.40

C494-500

  SDS
fisher scientific company
FLC494500
Poly Bottle 500g Each for $235.36
Case of 6 Each for $1,412.16
Description & Specifications

Specifications

Formula Weight 249.68
Melting Point 110°C
Quantity 2.5kg
DOT Information DOT Class 9, : Miscellaneous
Assay 98.5 to 100.5% (USP)
Molecular Formula CuO4S · 5 H2O
Physical Form Solid
Purity Grade USP
Vapor Pressure 7.3mmHg at 25°C
Color Blue
pH 3.5 to 4.5
Assay Percent Range (Dried Basis) 98.5to100.5%
Loss on Drying 33.0 to 36.5% (USP)
Calcium (Ca) 0.005% max. (USP)
Potassium (K) 0.01% max. (USP)
Synonym Copper (II) Sulfate Pentahydrate
Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Assay Percent Notes (Dried Basis) 98.5-100.5%
MDL Number MFCD00149681
Nickel (Ni) 0.005% max. (USP)
Sodium (Na) 0.02% max. (USP)
Identification Pass Test (USP)
Iron (Fe) 0.003% max. (USP)
Packaging Poly Bottle
Additional Information Vapor Pressure: 7.3mmHg at 25°C
Residual Solvents Meets Requirements (USP)
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1
ChemAlert Storage Symbol Blue

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1

WARNING!