Please login to your online account to display your discounted pricing

Cupric Sulfate Pentahydrate (Crystalline/Technical), Fisher Chemical


Quantity 10kg
Packaging Poly Pail

Manufacturer: fisher scientific company  FLC48910

 View more versions of this product

Catalog No. C489-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 249.68
Melting Point 110°C
Quantity 10kg
DOT Information DOT Class 9, : Miscellaneous
Physical Form Solid
Molecular Formula CuO4S · 5 H2O
Vapor Pressure 7.3mmHg at 25°C
Color Blue
pH 3.5 to 4.5
UN Number 3077
Identification Pass Test
CAS 7758-99-8
ChemAlert Storage Symbol Blue
Additional Information Vapor Pressure: 7.3mmHg at 25°C
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1
Chemical Name or Material Cupric Sulfate Pentahydrate
Packaging Poly Pail
MDL Number MFCD00149681
Synonym Copper (II) Sulfate
Health Hazard 2 WARNING!

Specifications

Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula CuO4S · 5 H2O
Formula Weight 249.68
MDL Number MFCD00149681
Synonym Copper (II) Sulfate
Accessories
 

Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1

WARNING!

SDS & Safety Documents