Please login to your online account to display your discounted pricing

Cupric Sulfate Pentahydrate, 98-102%, (Crystalline/ACS Reagent Grade), MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  0219141590

 View more versions of this product

Catalog No. ICN19141590

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range 98-102%
Molecular Formula CuSO4 · 5H2O
Formula Weight 249.68g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range 98-102%
Molecular Formula CuSO4 · 5H2O
Formula Weight 249.68g/mol
Accessories

Room Temperature