Please login to your online account to display your discounted pricing

Cupric Sulfate Pentahydrate, 98-102%, (Crystalline/ACS Reagent Grade), MP Biomedicals

$54.60 - $54.60

Chemical Identifiers

Chemical Name or Material Cupric Sulfate Pentahydrate
CAS 7758-99-8
Purity Grade ACS Reagent Grade
Physical Form Crystalline
Assay Percent Range 98-102%
Molecular Formula CuSO4 · 5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19141580

 
mp biomedicals
19141580
100g Each for $54.60
Description & Specifications

Specifications

Formula Weight 249.68
Purity Grade ACS Reagent Grade
Quantity 100g
Physical Form Crystalline
Chemical Name or Material Cupric Sulfate Pentahydrate
Assay Percent Range 98-102%
Molecular Formula CuSO4 · 5H2O
CAS 7758-99-8
Recommended Storage Room Temperature
Safety and Handling

Room Temperature