Please login to your online account to display your discounted pricing

Cupric Sulfate, Large Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$720.81 - $720.81

Chemical Identifiers

Chemical Name or Material Cupric Sulfate
CAS 7758-99-8
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CuSO4·5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002657

 
avantor performance
184105
Poly Bottle 2.5kg Each for $720.81
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 249.68
CAS 7758-99-8
Chemical Name or Material Cupric Sulfate
Molecular Formula CuSO4·5H2O
Packaging Poly Bottle
Physical Form Large Crystal
Specific Gravity 1L = 2.28kg
Quantity 2.5kg