Please login to your online account to display your discounted pricing

Cupric Sulfate, Fine Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$241.52 - $818.22

Chemical Identifiers

Chemical Name or Material Cupric Sulfate
CAS 7758-99-8
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CuSO4·5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002656

 
avantor performance
184319
Poly Bottle 1kg Each for $397.21

02002658

 
avantor performance
184305
Poly Bottle 2.5kg Each for $818.22

02002661

 
avantor performance
184301
Poly Bottle 500g Each for $241.52
Description & Specifications

Specifications

Formula Weight 249.68
Purity Grade Baker Analyzed™ ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Cupric Sulfate
CAS 7758-99-8
Molecular Formula CuSO4·5H2O
Specific Gravity 1L = 2.28kg
Quantity 1kg
Physical Form Fine Crystal