Please login to your online account to display your discounted pricing

Cupric Sulfate ACS AR Fine Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Cupric Sulfate
Name Note 5-Hydrate
CAS 7758-99-8
Purity Grade ACS Grade
Physical Form Fine Crystals
Molecular Formula CuSO4·5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4844212

  SDS
avantor performance
484406
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.68
Physical Form Fine Crystals
Purity Grade ACS Grade
Quantity 2.5kg
Packaging Poly
Name Note 5-Hydrate
CAS 7758-99-8
Chemical Name or Material Cupric Sulfate
Molecular Formula CuSO4·5H2O