Please login to your online account to display your discounted pricing

Cupric Sulfate, 5-Hydrate, U.S.P., J.T.Baker™

$301.65 - $1,046.77

Chemical Identifiers

Chemical Name or Material Cupric Sulfate
CAS 7758-99-8
Name Note 5-Hydrate
Purity Grade USP
Molecular Formula CuSO4·5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002659

 
avantor performance
184405
Wide Mouth Amber Glass Bottle 2.5kg Each for $1,046.77

02002662

 
avantor performance
184401
Wide Mouth Amber Glass Bottle 500g Each for $301.65
Description & Specifications

Specifications

Formula Weight 249.68
Packaging Wide Mouth Amber Glass Bottle
Quantity 2.5kg
Chemical Name or Material Cupric Sulfate
CAS 7758-99-8
Name Note 5-Hydrate
Purity Grade USP
Specific Gravity 1L = 2.28kg
Molecular Formula CuSO4·5H2O