missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cupric Citrate TS, Alkaline, Ricca Chemical

$228.73 - $420.82

Chemical Identifiers

CAS 7732-18-5,6132-04-3,497-19-8,7758-99-8
CAS Min % 76.0,13.15,7.6,1.31
CAS Max % 79.0,13.68,7.91,1.37
Physical Form Liquid
Chemical Name or Material Cupric Citrate, Test Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2493-16 Ricca Chemical Company
RI249316
Poly natural 500mL Each for $420.82
2493-4 Ricca Chemical Company
RI24934
Poly natural 120mL Each for $228.73
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6132-04-3,497-19-8,7758-99-8
79.0,13.68,7.91,1.37
Cupric Citrate, Test Solution
76.0,13.15,7.6,1.31
Liquid

Specifications

Liquid
500mL
76.0,7.6,1.31,13.15
Poly natural
Clear blue
79.0,7.91,1.37,13.68
7758-99-8,6132-04-3,7732-18-5,497-19-8
Cupric Citrate, Test Solution