Please login to your online account to display your discounted pricing

Cupric Chloride Dihydrate (Laboratory), Fisher Chemical


Quantity 3kg
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLC454-3

 View more versions of this product

Catalog No. C454-3

  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Identification Pass Test
Packaging Glass Bottle
Viscosity [NULL]
Color Blue Green
Melting Point 100°C / 212°F
pH 3.0 to 3.8
Quantity 3kg
Chemical Name or Material Cupric Chloride Dihydrate
CAS 10125-13-0
Purity Grade Laboratory
Physical Form Solid
Molecular Formula Cl2Cu · 2 H2O
Formula Weight 170.48g/mol
MDL Number MFCD00149674
Synonym Copper (II) Chloride
ChemAlert Storage Symbol White
Health Hazard 1 Signal Word:WARNING!
Health Hazard 3 Emergency Overview
Toxic if swallowed. Irritating to eyes, respiratory system and skin. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Corrosive to metal in aqueous
DOT Information DOT Class 8, : Corrosive

Specifications

Chemical Name or Material Cupric Chloride Dihydrate
CAS 10125-13-0
Purity Grade Laboratory
Physical Form Solid
Molecular Formula Cl2Cu · 2 H2O
Formula Weight 170.48g/mol
MDL Number MFCD00149674
Synonym Copper (II) Chloride
Accessories
 

Signal Word:WARNING!

Emergency Overview
Toxic if swallowed. Irritating to eyes, respiratory system and skin. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Corrosive to metal in aqueous

SDS & Safety Documents