Please login to your online account to display your discounted pricing

Cupric Chloride Dihydrate (Laboratory), Fisher Chemical

$214.43 - $214.43

Chemical Identifiers

Chemical Name or Material Cupric Chloride Dihydrate
CAS 10125-13-0
Purity Grade Laboratory Grade
Physical Form Solid
Molecular Formula Cl2Cu · 2 H2O
Formula Weight 170.48
MDL Number MFCD00149674
Synonym Copper (II) Chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

C454-500

  SDS
fisher scientific company
FLC454500
Glass Bottle 500g Each for $214.43
Case of 6 Each for $1,286.58
Description & Specifications

Specifications

Purity Grade Laboratory Grade
Formula Weight 170.48
Melting Point 100°C
Quantity 500g
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula Cl2Cu · 2 H2O
Viscosity [NULL]
Color Blue Green
pH 3.0 to 3.8
Identification Pass Test
Chemical Name or Material Cupric Chloride Dihydrate
Packaging Glass Bottle
MDL Number MFCD00149674
CAS 10125-13-0
ChemAlert Storage Symbol White
Synonym Copper (II) Chloride
Health Hazard 1 Signal Word:WARNING!
Health Hazard 3 Emergency Overview
Toxic if swallowed. Irritating to eyes, respiratory system and skin. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Corrosive to metal in aqueous