Please login to your online account to display your discounted pricing

Cupric Chloride, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$259.04 - $2,387.20

Chemical Identifiers

Chemical Name or Material Cupric Chloride
CAS 10125-13-0
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CuCl2·2H2O
Formula Weight 170.48
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002651

 
avantor performance
179204
Poly Bottle 125g Each for $259.04

02002652

 
avantor performance
179205
Poly Bottle 2.5kg Each for $2,387.20

02002653

 
avantor performance
179201
Poly Bottle 500g Each for $623.34
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 170.48
Chemical Name or Material Cupric Chloride
Molecular Formula CuCl2·2H2O
CAS 10125-13-0
Name Note Dihydrate
Packaging Poly Bottle
Specific Gravity 1L = 2.54kg
Quantity 125g