Please login to your online account to display your discounted pricing

Cupric Acetate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$361.39 - $1,089.75

Chemical Identifiers

Chemical Name or Material Cupric Acetate
CAS 6046-93-1
Name Note Monohydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3COO)2Cu·H2O
Formula Weight 199.65
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002648

 
avantor performance
176605
Poly Bottle 2.5kg Each for $1,089.75

02002649

 
avantor performance
176601
Poly Bottle 500g Each for $361.39
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 199.65
Chemical Name or Material Cupric Acetate
Name Note Monohydrate
Packaging Poly Bottle
Specific Gravity 1L = 1.88kg
Molecular Formula (CH3COO)2Cu·H2O
CAS 6046-93-1
Quantity 2.5kg