Please login to your online account to display your discounted pricing

Cupric Acetate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  176601

 View more versions of this product

Catalog No. 02002649

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 199.65
Chemical Name or Material Cupric Acetate
CAS 6046-93-1
Molecular Formula (CH3COO)2Cu·H2O
Name Note Monohydrate
Packaging Poly Bottle
Specific Gravity 1L = 1.88kg
Quantity 500g

Specifications

Chemical Name or Material Cupric Acetate
CAS 6046-93-1
Name Note Monohydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3COO)2Cu·H2O
Formula Weight 199.65