Please login to your online account to display your discounted pricing

Cupferron, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC423600250

 View more versions of this product

Catalog No. AC423600250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 155.16
Quantity 25g
TSCA TSCA
RTECS Number NC4725000
EINECS Number 205-183-2
Synonym N-Hydroxy-N-nitrosobenzenamine ammonium salt
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H9N3O2
Chemical Name or Material Cupferron
Linear Formula C6H4N(ONH4)NO
MDL Number MFCD00078422
CAS 135-20-6
Health Hazard 1 CANCER SUSPECTED/HYGROSCOPIC

Specifications

Chemical Name or Material Cupferron
CAS 135-20-6
Purity Grade ACS Reagent Grade
Molecular Formula C6H9N3O2
Linear Formula C6H4N(ONH4)NO
Formula Weight 155.16
MDL Number MFCD00078422
Synonym N-Hydroxy-N-nitrosobenzenamine ammonium salt
Accessories

CANCER SUSPECTED/HYGROSCOPIC

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.
Causes skin irritation.

SDS & Safety Documents