Please login to your online account to display your discounted pricing

Alfa Aesar™ Cupferron, 97+%

Chemical Identifiers

Chemical Name or Material Cupferron
CAS 135-20-6
Molecular Formula C6H9N3O2
Formula Weight 155.16
MDL Number MFCD00078422
UN Number UN2811
Merck Index 14,2622
Synonym N-Nitrosophenylhydroxylamine ammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1655114

 
alfa aesar™
A1655114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.16
Chemical Name or Material Cupferron
TSCA TSCA
Odor Odorless
Assay ≥97%
Molecular Formula C6H9N3O2
MDL Number MFCD00078422
CAS 135-20-6
UN Number UN2811
Merck Index 14,2622
Synonym N-Nitrosophenylhydroxylamine ammonium salt
Health Hazard 1 Danger
Melting Point 154°C (dec.)
Health Hazard 2 GHS H Statement
H301-H351-H315-H319-H317
Toxic if swallowed.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number NC4725000
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H351-H315-H319-H317
Toxic if swallowed.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

2° to 8°C