Please login to your online account to display your discounted pricing

R&D Systems Cultrex Rat Collagen I (LV), Lower Viscosity

$39.11 - $195.54

Specifications

Concentration 3mg/mL
Source Rat tail tendons
Endotoxin Concentration <20EU/mL
For Use With (Application) For research use only
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

344300301

 
r&d systems
344300301
Each for $39.11

344310001

 
r&d systems
344310001
Each for $195.54
Description & Specifications

Specifications

Concentration 3mg/mL
Source Rat tail tendons
Endotoxin Concentration <20EU/mL
For Use With (Application) For research use only