Please login to your online account to display your discounted pricing

R&D Systems Cultrex PathClear Reduced Growth Factor BME, Type 3, with Phenol Red

Physiologically aligned with the in vivo solid tumors environment and is recommended for xenografts and other in vivo applications

$191.40 - $287.11

Specifications

Concentration 12-18mg/mL
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

36-320-0502P

 
r&d systems
363200502P
5mL Each for $191.40

36-320-1002P

 
r&d systems
363201002P
2x5mL Each for $287.11
Description & Specifications

Specifications

Concentration 12-18mg/mL

Basement membranes are continuous sheets of specialized extracellular matrix that form an interface between endothelial, epithelial, muscle, or neuronal cells and their adjacent stroma. Basement membranes are degraded and regenerated during development and wound healing. They not only support cells and cell layers, but they also play an essential role in tissue organization that affects cell adhesion, migration, proliferation, and differentiation. Basement membranes provide major barriers to invasion by metastatic tumor cells.

The major components of BME include laminin, collagen IV, entactin, and heparin sulfate proteoglycan
BME can be used in a multiple applications, under a variety of cell culture conditions, for maintaining growth or promoting differentiation of primary endothelial, epithelial, smooth muscle and stem cells
BME can also be utilized in cell attachment, neurite outgrowth, angiogenesis, in vitro cell invasion and in vivo tumorigenicity assays
Higher tensile strength than standard BME
RPMI1640 medium without phenol red
Sterility tested

Applications:

solid tumors