Please login to your online account to display your discounted pricing

R&D Systems Cultrex Basement Membrane Extract, PathClear With Phenol Red

$195.54 - $293.30

Specifications

Concentration 12 to 18mg/mL
Concentration 12 to 18mg/mL
Source Murine Engelbreth-Holm-Swarm tumor
Endotoxin Concentration ≤8EU/mL
For Use With (Application) Can be used for promotion and maintenance of a differentiated phenotype in a variety of cell cultures including primary epithelial cells, endothelial cells, and smooth muscle cells
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

343200501P

 
r&d systems
343200501P
Each for $195.54

343201001P

 
r&d systems
343201001P
Each for $293.30
Description & Specifications

Specifications

Concentration 12 to 18mg/mL
Concentration 12 to 18mg/mL
Source Murine Engelbreth-Holm-Swarm tumor
Endotoxin Concentration ≤8EU/mL
For Use With (Application) Can be used for promotion and maintenance of a differentiated phenotype in a variety of cell cultures including primary epithelial cells, endothelial cells, and smooth muscle cells